Läs mer
4-[2,2-Dichloro-1-(4-hydroxyphenyl)ethenyl]phenol, TRC
Produktkod. 30135851
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135851 100 mg
30135603 500 mg
30003811 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135851 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD438908100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14868-03-2
Molekylformel C14 H10 Cl2 O2
Molekylvikt (g/mol) 281.134
Synonym Phenol, 4,4'-(dichloroethenylidene)bis- (9CI), Phenol, 4,4'-(dichlorovinylidene)di- (6CI,8CI), 4,4'-(2,2-Dichloroethenylidene)bis[phenol], 1,1-Bis(4-hydroxyphenyl)-2,2-dichloroethylene, 1,1-Bis(p-hydroxyphenyl)-2,2-dichloroethylene, 1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethene, 1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene, 1,1-Dichloro-2,2-bis(p-hydroxyphenyl)ethylene, 2,2-Bis(4-hydroxyphenyl)-1,1-dichloroethene, 2,2-Bis(4-hydroxyphenyl)-1,1-dichloroethylene, 2,2-Dichloro-1,1-bis(4-hydroxyphenyl)ethylene, 4,4'-(Dichlorovinylidene)diphenol, Bis(4-hydroxyphenyl)-2,2-dichloroethylene, Bis(4-hydroxyphenyl)dichloroethylene, Bisphenol C 2
IUPAC-namn 4-[2,2-dichloro-1-(4-hydroxyphenyl)ethenyl]phenol
LEDER Oc1ccc(cc1)C(=C(Cl)Cl)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material Bisphenol C 2
Kvantitet 100 mg
InChI formel InChI=1S/C14H10Cl2O2/c15-14(16)13(9-1-5-11(17)6-2-9)10-3-7-12(18)8-4-10/h1-8,17-18H
Formel vikt 280.0058