Läs mer
4-(2,3-Epoxypropoxy)phenylacetic Acid, Mikromol™
Produktkod. 30050305
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050305 100 mg
30052623 25 mg
30459126 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30050305 Leverantör Mikromol Leverantörsnummer MM0013.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(2,3-Epoxypropoxy)phenylacetic Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(2,3-Epoxypropoxy)phenylacetic Acid
CAS 72224-25-0
Analyt- eller komponentnamn 4-(2,3-Epoxypropoxy)phenylacetic Acid
Frakt skick Ice pack (-20°C)
Molekylformel C11 H12 O4
Certifikat och efterlevnad ISO 9001
Synonym 4-(2-oxiranylmethoxy)-Benzeneacetic acid, Benzeneacetic acid, 4-(oxiranylmethoxy)- (9CI), Benzeneacetic acid, 4-(oxiranylmethoxy)-, (+/-)-
InChI formel InChI=1S/C11H12O4/c12-11(13)5-8-1-3-9(4-2-8)14-6-10-7-15-10/h1-4,10H,5-7H2,(H,12,13)
LEDER OC(=O)Cc1ccc(OCC2CO2)cc1
IUPAC-namn 2-[4-(oxiran-2-ylmethoxy)phenyl]acetic acid
Molekylvikt (g/mol) 208.21
Kvantitet 100 mg
Formel vikt 208.0736 g/mol