Läs mer
4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(dimethylamino)propoxy)-6-methoxyquinoline-3-carbonitrile Dimer, TRC
Produktkod. 30377196
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30377196 10 mg
30375593 100 mg
30375771 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30377196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD42457010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2095306-26-4
Molekylformel C46H44Cl4N8O6
Molekylvikt (g/mol) 946.7
Synonym 4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(4-(3-((4-((2,4-dichloro-5-methoxyphenyl)amino)-3-isocyano-6-methoxyquinolin-7-yl)oxy)propyl)piperazin-1-yl)propoxy)-6-methoxyquinoline-3-carbonitrile, Bosutinib dimer
LEDER COc1cc(Nc2c(cnc3cc(OCCCN4CCN(CCCOc5cc6ncc([N+]#[C-])c(Nc7cc(OC)c(Cl)cc7Cl)c6cc5OC)CC4)c(OC)cc23)C#N)c(Cl)cc1Cl
Specifikationer
Kemiskt namn eller material 4-((2,4-Dichloro-5-methoxyphenyl)amino)-7-(3-(dimethylamino)propoxy)-6-methoxyquinoline-3-carbonitrile Dimer
Kvantitet 10 mg
InChI formel InChI=1S/C46H44Cl4N8O6/c1-52-38-26-54-35-21-44(42(62-5)17-29(35)46(38)56-37-23-40(60-3)33(50)19-31(37)48)64-15-7-9-58-12-10-57(11-13-58)8-6-14-63-43-20-34-28(16-41(43)61-4)45(27(24-51)25-53-34)55-36-22-39(59-2)32(49)18-30(36)47/h16-23,25-26H,6-15H2,2-5H3,(H,53,55)(H,54,56)
Formel vikt 944.2138