missing translation for 'onlineSavingsMsg'
Learn More

4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7053615
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
119516-86-8
C13H12N2
196.253
FNDFKQYZEDOHRC-UHFFFAOYSA-N
1-4-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethylpyrrol-1-yl benzonitrile, 4-2,5-dimethyl-1h-pyrrol-1-yl benzonitrile, 2,5-dimethyl-1-4-cyanophenyl pyrrole, n-4'-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethyl-1-pyrrolyl benzonitrile, benzonitrile,4-2,5-dimethyl-1h-pyrrol-1-yl
3787857
4-(2,5-dimethylpyrrol-1-yl)benzonitrile
CC1=CC=C(N1C2=CC=C(C=C2)C#N)C
This item is not returnable. View return policy
119516-86-8
C13H12N2
196.253
FNDFKQYZEDOHRC-UHFFFAOYSA-N
1-4-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethylpyrrol-1-yl benzonitrile, 4-2,5-dimethyl-1h-pyrrol-1-yl benzonitrile, 2,5-dimethyl-1-4-cyanophenyl pyrrole, n-4'-cyanophenyl-2,5-dimethylpyrrole, 4-2,5-dimethyl-1-pyrrolyl benzonitrile, benzonitrile,4-2,5-dimethyl-1h-pyrrol-1-yl
3787857
4-(2,5-dimethylpyrrol-1-yl)benzonitrile
CC1=CC=C(N1C2=CC=C(C=C2)C#N)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN3439
Formula Weight 196.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'