Läs mer
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzonitrile, TRC
Produktkod. 30387849
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30387849 500 mg
30371388 100 mg
30375436 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30387849 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD496120500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31489-18-6
Molekylformel C11H6N2O2
Molekylvikt (g/mol) 198.17
Synonym N-(4-Cyanophenyl)maleimide, N-(p-Cyanophenyl)maleimide
LEDER O=C1C=CC(=O)N1c2ccc(cc2)C#N
Specifikationer
Kemiskt namn eller material 4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzonitrile
Kvantitet 500 mg
InChI formel InChI=1S/C11H6N2O2/c12-7-8-1-3-9(4-2-8)13-10(14)5-6-11(13)15/h1-6H
Formel vikt 198.0429