Läs mer
4-[2’-(7’’-Deazaguanine)ethyl]benzoic Acid N-Hydroxysuccinimide Ester, TRC
Produktkod. 30351487
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30351487 10 mg
1 options
Produktkod. 30351487 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD20135010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2'-(7''-Deazaguanine)ethyl]benzoic Acid N-Hydroxysuccinimide Ester
CAS 204257-65-8
Molekylformel C19 H17 N5 O5
Kvantitet 10 mg
Synonym Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, 2,5-dioxo-1-pyrrolidinyl ester, 2,5-Pyrrolidinedione, 1-[[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]oxy]- (9CI), 4-[2'-(7''-Deazaguanine)ethyl]benzoic acid N-hydroxysuccinimide ester
InChI formel InChI=1S/C19H17N5O5/c20-19-22-16-15(17(27)23-19)12(9-21-16)6-3-10-1-4-11(5-2-10)18(28)29-24-13(25)7-8-14(24)26/h1-2,4-5,9H,3,6-8H2,(H4,20,21,22,23,27)
LEDER NC1=NC(=O)c2c(CCc3ccc(cc3)C(=O)ON4C(=O)CCC4=O)c[nH]c2N1
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate
Molekylvikt (g/mol) 395.37
Formel vikt 395.123