Läs mer
4-(2-Acetoxy-ethyl)phenyl Acetate, TRC
Produktkod. 30326994
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30326994 5 g
1 options
Produktkod. 30326994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1656005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(2-Acetoxy-ethyl)phenyl Acetate
CAS 60037-42-5
Molekylformel C12H14O4
Kvantitet 5 g
Synonym 2-(4-Acetoxyphenyl)ethyl Acetate, 2-(p-Acetoxyphenyl)ethyl Acetate, 4-(2-Acetoxyethyl)phenyl Acetate, Acetic Acid 4-(2-acetoxyethyl)phenyl Ester, Diacetyl-1,4-tyrosol
InChI formel InChI=1S/C12H14O4/c1-9(13)15-8-7-11-3-5-12(6-4-11)16-10(2)14/h3-6H,7-8H2,1-2H3
LEDER CC(=O)OCCc1ccc(OC(=O)C)cc1
IUPAC-namn 2-(4-acetyloxyphenyl)ethyl acetate
Molekylvikt (g/mol) 222.24
Formel vikt 222.0892