Läs mer
4-(2-amino-2-methylpropyl)phenol, TRC
Produktkod. 30134902
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134902 250 mg
30004934 100 mg
30010233 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134902 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA620088250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51706-55-9
Molekylformel C10 H15 N O
Molekylvikt (g/mol) 165.232
Synonym 4-(2-Amino-2,2-dimethylethyl)Phenol, 4-(2-Methyl-2-Aminopropyl)Phenol, p-Hydroxyphentermine
IUPAC-namn 4-(2-amino-2-methyl-propyl)phenol
LEDER CC(C)(N)Cc1ccc(O)cc1
Specifikationer
Kemiskt namn eller material 4-(2-amino-2-methylpropyl)phenol
Kvantitet 250 mg
InChI formel InChI=1S/C10H15NO/c1-10(2,11)7-8-3-5-9(12)6-4-8/h3-6,12H,7,11H2,1-2H3
Formel vikt 165.1154