missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Bromo-6-fluorobenzyl)morpholine, 96%

Brand:  Alfa Aesar™ H62229.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468778

  • 2082.56 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(2-Bromo-6-fluorobenzyl)morpholine
274.13

Specifications

4-(2-Bromo-6-fluorobenzyl)morpholine
96%
274.13
Solid
1g
Videos
SDS