Läs mer
4-(2-Bromoacetamido)-TEMPO, TRC
Produktkod. 30240640
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240640 25 mg
30240655 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB67906825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24567-97-3
Molekylformel C11H20BrN2O2
Molekylvikt (g/mol) 292.19
Synonym 4-[(2-Bromoacetyl)amino]-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-(2-Bromoacetamido)-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-(2-Bromoacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-?-Bromoacetamido-2,2,6,6-tetramethylpiperidino-N-oxy, N-(1-Oxyl-2,2,6,6-tetramethyl-4-piperidinyl)bromoacetamide, N-(1-Oxyl-2,2,6,6-tetramethyl-4-piperidyl)bromoacetamide, 4-(2-Bromoacetamido)-2,2,6,6-tetramethyl-piperidinooxy, 4-[(Bromoacetyl)amino]-2,2,6,6-tetramethyl-1-piperidinyloxy
IUPAC-namn 2-bromo-N-(1-hydroxy-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
LEDER [O]N1C(C)(C)CC(NC(CBr)=O)CC1(C)C
Specifikationer
Kemiskt namn eller material 4-(2-Bromoacetamido)-TEMPO
Kvantitet 25 mg
InChI formel InChI=1S/C11H20BrN2O2/c1-10(2)5-8(13-9(15)7-12)6-11(3,4)14(10)16/h8H,5-7H2,1-4H3,(H,13,15)
Formel vikt 291.07