Läs mer
4-(2-Carboxyethenyl)benzoic Acid Methyl Ester, TRC
Produktkod. 30361431
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30361431 2.5 g
1 options
Produktkod. 30361431 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1780952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(2-Carboxyethenyl)benzoic Acid Methyl Ester
CAS 19473-96-2
Molekylformel C11H10O4
Kvantitet 2.5 g
Synonym p-Carbomethoxycinnamic Acid, p-Methoxycarbonylcinnamic Acid
InChI formel InChI=1S/C11H10O4/c1-15-11(14)9-5-2-8(3-6-9)4-7-10(12)13/h2-7H,1H3,(H,12,13)/b7-4+
LEDER COC(=O)c1ccc(\C=C\C(=O)O)cc1
IUPAC-namn (E)-3-(4-methoxycarbonylphenyl)prop-2-enoic acid
Molekylvikt (g/mol) 206.19
Formel vikt 206.0579