Learn More
4-(2-Carboxyethenyl)benzoic Acid Methyl Ester, TRC
Product Code. 30349136
Change view
Click to view available options
Kvantitet:
250 mg
Product Code. Kvantitet
30349136 250 mg
1 options
Product Code. 30349136 Supplier Toronto Research Chemicals Supplier No. TRCC178095250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-(2-Carboxyethenyl)benzoic Acid Methyl Ester
CAS 19473-96-2
Molekylformel C11H10O4
Kvantitet 250 mg
Synonym p-Carbomethoxycinnamic Acid, p-Methoxycarbonylcinnamic Acid
InChI formel InChI=1S/C11H10O4/c1-15-11(14)9-5-2-8(3-6-9)4-7-10(12)13/h2-7H,1H3,(H,12,13)/b7-4+
LEDER COC(=O)c1ccc(\C=C\C(=O)O)cc1
IUPAC-namn (E)-3-(4-methoxycarbonylphenyl)prop-2-enoic acid
Molekylvikt (g/mol) 206.19
Formel vikt 206.0579