missing translation for 'onlineSavingsMsg'
Learn More

4'-(2-Chloroethyl)acetophenone, tech. 90%, Thermo Scientific™

Catalog Number p-7046285
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
69614-95-5
C10H11ClO
182.647
MFCD00016350
RGVUACHOYCYNMB-UHFFFAOYSA-N
4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
144359
1-[4-(2-chloroethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)CCCl
This item is not returnable. View return policy
69614-95-5
C10H11ClO
182.647
MFCD00016350
RGVUACHOYCYNMB-UHFFFAOYSA-N
4'-beta-chloroethyl acetophenone, 4-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethanone, 4'-2-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl-1-ethanone, 4'-.beta.-chloroethyl acetophenone, p-beta-chloroethyl acetophenone, 1-4-2-chloroethyl phenyl ethan-1-one, 1-acetyl-4-2-chloroethyl benzene
144359
1-[4-(2-chloroethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)CCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.15
Boiling Point 112°C (0.1 mmHg)
Refractive Index 1.555
Quantity 5 g
Beilstein 2081019
Formula Weight 182.65
Percent Purity 90%
Grade Technical
Chemical Name or Material 4'-(2-Chloroethyl)acetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'