Läs mer
4-[2-(Dimethylamino)ethoxy]benzonitrile, TRC
Produktkod. 30134830
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134830 250 mg
30134151 100 mg
30007825 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD462100250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24197-95-3
Molekylformel C11H14N2O
Molekylvikt (g/mol) 190.242
Synonym p-[2-(Dimethylamino)ethoxy]-benzonitrile;
IUPAC-namn 4-[2-(dimethylamino)ethoxy]benzonitrile
LEDER CN(C)CCOc1ccc(cc1)C#N
Specifikationer
Kemiskt namn eller material 4-[2-(Dimethylamino)ethoxy]benzonitrile
Kvantitet 250 mg
InChI formel InChI=1S/C11H14N2O/c1-13(2)7-8-14-11-5-3-10(9-12)4-6-11/h3-6H,7-8H2,1-2H3
Formel vikt 190.1106