Läs mer
4-(2-Ethoxybenzamido)-1-methyl-3-n-propylpyrazole-5-carboxamide, TRC
Produktkod. 30373730
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373730 100 mg
30379119 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30373730 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE891000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139756-03-9
Molekylformel C17 H22 N4 O3
Molekylvikt (g/mol) 330.38
Synonym 1H-Pyrazole-5-carboxamide, 4-[(2-ethoxybenzoyl)amino]-1-methyl-3-propyl-, 4-[(2-Ethoxybenzoyl)amino]-1-methyl-3-propyl-1H-pyrazole-5-carboxamide, 4-(2-Ethoxybenzamido)-1-methyl-3-propylpyrazole-5-carboxamide, 4-(2-Ethoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
IUPAC-namn 4-[(2-ethoxybenzoyl)amino]-2-methyl-5-propylpyrazole-3-carboxamide
LEDER CCCc1nn(C)c(C(=O)N)c1NC(=O)c2ccccc2OCC
Specifikationer
Kemiskt namn eller material 4-(2-Ethoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
Kvantitet 100 mg
InChI formel InChI=1S/C17H22N4O3/c1-4-8-12-14(15(16(18)22)21(3)20-12)19-17(23)11-9-6-7-10-13(11)24-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H2,18,22)(H,19,23)
Formel vikt 330.1692