missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Fluoro-4-nitrophenyl)morpholine, 98%

Brand:  Alfa Aesar™ H61940.06

Additional Details : CAS Number : 2689-39-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15448528

  • 253.57 SEK / 5g


Estimated Shipment date
from Supplier 14-11-2018
Add to basket

Specifications

Chemical Identifiers

2689-39-6
226.207
ZEQCFSSBZPZEFJ-UHFFFAOYSA-N
2905489
C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])F
C10H11FN2O3
MFCD03138381
4-2-fluoro-4-nitrophenyl morpholine, morpholine, 4-2-fluoro-4-nitrophenyl, 4-2-fluoro-4-nitro-phenyl-morpholine, 3-fluoro-4-morpholinonitrobenzene, acmc-209gtt, cambridge id 6614516, 3fluoro-4-morpholinonitrobenzene, 3-fluoro-4-morpholinyl nitrobenzene, 3-fluoro-4-morpholinyl-nitrobenzene, 1-fluoro-2-morpholino-5-nitrobenzene
4-(2-fluoro-4-nitrophenyl)morpholine

Specifications

4-(2-Fluoro-4-nitrophenyl)morpholine
2689-39-6
MFCD03138381
ZEQCFSSBZPZEFJ-UHFFFAOYSA-N
4-(2-fluoro-4-nitrophenyl)morpholine
2905489
Solid
5g
C10H11FN2O3
4-2-fluoro-4-nitrophenyl morpholine, morpholine, 4-2-fluoro-4-nitrophenyl, 4-2-fluoro-4-nitro-phenyl-morpholine, 3-fluoro-4-morpholinonitrobenzene, acmc-209gtt, cambridge id 6614516, 3fluoro-4-morpholinonitrobenzene, 3-fluoro-4-morpholinyl nitrobenzene, 3-fluoro-4-morpholinyl-nitrobenzene, 1-fluoro-2-morpholino-5-nitrobenzene
C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])F
226.207
226.2
98%
Videos
SDS