missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Fluoro-6-nitrophenyl)morpholine, 97%

Brand:  Alfa Aesar™ H63272.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449888

  • 762.64 SEK / 1g


Estimated Shipment date
from Supplier 16-11-2018
Add to basket

Specifications

Chemical Identifiers

1233955-40-2
226.207
RVPOFXBMBJKJJK-UHFFFAOYSA-N
53255510
C1COCCN1C2=C(C=CC=C2F)[N+](=O)[O-]
C10H11FN2O3
MFCD13689260
4-2-fluoro-6-nitrophenyl morpholine
4-(2-fluoro-6-nitrophenyl)morpholine

Specifications

4-(2-Fluoro-6-nitrophenyl)morpholine
1233955-40-2
MFCD13689260
RVPOFXBMBJKJJK-UHFFFAOYSA-N
4-(2-fluoro-6-nitrophenyl)morpholine
53255510
Solid
1g
C10H11FN2O3
4-2-fluoro-6-nitrophenyl morpholine
C1COCCN1C2=C(C=CC=C2F)[N+](=O)[O-]
226.207
226.2
97%
Videos
SDS