Läs mer
4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde, TRC
Produktkod. 30007066
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007066 100 mg
30006138 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH946735100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29122-74-5
Molekylformel C13H19NO3
Molekylvikt (g/mol) 237.29
Synonym 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde, p-[2-Hydroxy-3-(isopropylamino)propoxy]benzaldehyde (8CI), H 128/80, 4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde, Metoprolol Tartrate Imp. C (EP)
IUPAC-namn 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]benzaldehyde
LEDER CC(C)NCC(O)COc1ccc(C=O)cc1
Specifikationer
Kemiskt namn eller material 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H19NO3/c1-10(2)14-7-12(16)9-17-13-5-3-11(8-15)4-6-13/h3-6,8,10,12,14,16H,7,9H2,1-2H3
Formel vikt 237.1365
Procent renhet >95%