Läs mer
4-(2-Hydroxy-3-isopropylaminopropoxy)benzoic Acid, TRC
Produktkod. 30012836
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012836 50 mg
30011339 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012836 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94357550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72570-70-8
Molekylformel C13 H19 N O4
Molekylvikt (g/mol) 253.29
Synonym Benzoic acid, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, 4-[2-hydroxy-3-(isopropylamino)propoxy]benzoic acid
IUPAC-namn 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]benzoic acid
LEDER CC(C)NCC(O)COc1ccc(cc1)C(=O)O
Specifikationer
Kemiskt namn eller material 4-(2-Hydroxy-3-isopropylaminopropoxy)benzoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C13H19NO4/c1-9(2)14-7-11(15)8-18-12-5-3-10(4-6-12)13(16)17/h3-6,9,11,14-15H,7-8H2,1-2H3,(H,16,17)
Formel vikt 253.1314