missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyethyl)benzonitrile, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Spécification
Assay Percent Range | 97% |
Linear Formula | CNC6H4(CH2)2OH |
Quantity | 250 mg |
Formula Weight | 147.18 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Hydroxyethyl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'