Learn More
4-(2-Iodophenoxy)tetrahydropyran, 97%, Thermo Scientific™
Product Code. 10776725
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10776725 1 g Amber Glass Bottle 1g
10387154 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10776725 Supplier Thermo Scientific Maybridge Supplier No. CC52211DA
 Restricted Item
Beräknad leverans: 14-12-2026
Add to basket
Chemical Identifiers
CAS 906352-70-3
Molecular Formula C11H13IO2
Molecular Weight (g/mol) 304.127
MDL Number MFCD09064961
InChI Key ADTBIBCNKWWYRD-UHFFFAOYSA-N
Synonym 4-2-iodophenoxy tetrahydropyran, 4-2-iodophenoxy oxane, 4-2-iodophenoxy tetrahydro-2h-pyran
PubChem CID 21981312
IUPAC Name 4-(2-iodophenoxy)oxane
SMILES C1COCCC1OC2=CC=CC=C2I
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 304.12
Percent Purity 97%
Chemical Name or Material 4-(2-Iodophenoxy)tetrahydropyran