Läs mer
4-[(2-Isopropoxyethoxy)methyl]phenol, TRC
Produktkod. 30138680
Change view
Kvantitet:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138680 1 g
30007613 250 mg
30010412 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138680 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI8233001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 177034-57-0
Molekylformel C12 H18 O3
Molekylvikt (g/mol) 210.27
Synonym 4-[(2-Isopropoxyethoxy)methyl]phenol, Bisoprolol Fumarate Imp. M (Pharmeuropa)
IUPAC-namn 4-(2-propan-2-yloxyethoxymethyl)phenol
LEDER CC(C)OCCOCc1ccc(O)cc1
Specifikationer
Kemiskt namn eller material 4-[(2-Isopropoxyethoxy)methyl]phenol
Kvantitet 1 g
InChI formel InChI=1S/C12H18O3/c1-10(2)15-8-7-14-9-11-3-5-12(13)6-4-11/h3-6,10,13H,7-9H2,1-2H3
Formel vikt 210.1256