Läs mer
4-(2-Methyl-1-pyrrolidinyl)-7-chloroquinoline, TRC
Produktkod. 30133756
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133756 10 mg
30011873 5 mg
30011377 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36999010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6281-58-9
Molekylformel C14H15ClN2
Molekylvikt (g/mol) 246.74
Synonym 7-Chloro-4-(2-methyl-1-pyrrolidinyl)-quinoline
IUPAC-namn 7-chloro-4-(2-methylpyrrolidin-1-yl)quinoline
LEDER ClC1=CC=C2C(N3C(C)CCC3)=CC=NC2=C1
Specifikationer
Kemiskt namn eller material 4-(2-Methyl-1-pyrrolidinyl)-7-chloroquinoline
Kvantitet 10 mg
InChI formel InChI=1S/C14H15ClN2/c1-10-3-2-8-17(10)14-6-7-16-13-9-11(15)4-5-12(13)14/h4-7,9-10H,2-3,8H2,1H3
Formel vikt 246.0924