Läs mer
4-[2-(Methyl-2-pyridinylamino)ethoxy]benzenepropanamide(Rosiglitazone Impurity), TRC
Produktkod. 30346521
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30346521 100 mg
1 options
Produktkod. 30346521 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB288665100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanamide
CAS 1797979-55-5
Molekylformel C17 H21 N3 O2
Kvantitet 100 mg
Synonym 3-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanamide, Rosiglitazone Maleate Imp. A (Pharmeuropa), 4-[2-(Methyl-2-pyridinylamino)ethoxy]benzenepropanamide (ACI), Benzenepropanamide, 4-[2-(methyl-2-pyridinylamino)ethoxy]- (ACI), 3-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanamide, Rosiglitazone Maleate Imp. A (Pharmeuropa), Rosiglitazone maleate related, SB-211656, SB214521, SB211656, SB236833, SB237216, Rosiglitazone Maleate Impurity A, Rosiglitazone Impurity A
InChI formel InChI=1S/C17H21N3O2/c1-20(17-4-2-3-11-19-17)12-13-22-15-8-5-14(6-9-15)7-10-16(18)21/h2-6,8-9,11H,7,10,12-13H2,1H3,(H2,18,21)
LEDER CN(CCOc1ccc(CCC(=O)N)cc1)c2ccccn2
IUPAC-namn 3-[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanamide
Molekylvikt (g/mol) 299.37
Formel vikt 299.1634