missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Methyl-2-pyridinylamino)ethoxy]benzenepropanoic Acid Methyl Ester, TRC

Product Code. 30481518
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30481518 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481518 Supplier Toronto Research Chemicals Supplier No. TRCM328215100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-[2-(Methyl-2-pyridinylamino)ethoxy]benzenepropanoic Acid Methyl Ester
CAS 153939-51-6
Molekylformel C31 H41 N O12
Kvantitet 100 mg
InChI formel InChI=1S/C31H41NO12/c1-5-39-25(36)16-10-8-6-7-9-13-17-40-31-26(32-29(37)22-14-11-12-15-23(22)30(32)38)28(43-21(4)35)27(42-20(3)34)24(44-31)18-41-19(2)33/h11-12,14-15,24,26-28,31H,5-10,13,16-18H2,1-4H3/t24?,26?,27-,28?,31+/m0/s1
LEDER CCOC(=O)CCCCCCCCO[C@@H]1OC(COC(=O)C)[C@H](OC(=O)C)C(OC(=O)C)C1N2C(=O)c3ccccc3C2=O
IUPAC-namn ethyl 9-[(2R,5R)-4,5-diacetyloxy-6-(acetyloxymethyl)-3-(1,3-dioxoisoindol-2-yl)oxan-2-yl]oxynonanoate
Molekylvikt (g/mol) 619.657
Formel vikt 619.263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.