missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide, Alfa Aesar™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 250 mg |
Formula Weight | 292.36 |
Chemical Name or Material | 4-[2-(Methylaminomethyl)phenoxy]benzenesulfonamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'