missing translation for 'onlineSavingsMsg'
Läs mer

4-(2-Methylpropyl)-2-oxo-1-pyrrolidineacetic Acid, TRC

Produktkod. 30464871
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464871 10 mg
30476284 100 mg
30473177 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30464871 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM70480010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1511511-44-6
Molekylformel C10H17NO3
Molekylvikt (g/mol) 199.25
Synonym Pregabalin Impurity, 2-(4-Isobutyl-2-oxopyrrolidin-1-yl)acetic Acid
IUPAC-namn 2-[4-(2-methylpropyl)-2-oxopyrrolidin-1-yl]acetic acid
LEDER CC(C)CC1CN(CC(=O)O)C(=O)C1

Specifications

Kemiskt namn eller material 4-(2-Methylpropyl)-2-oxo-1-pyrrolidineacetic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C10H17NO3/c1-7(2)3-8-4-9(12)11(5-8)6-10(13)14/h7-8H,3-6H2,1-2H3,(H,13,14)
Formel vikt 199.1208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.