Läs mer
4-(2-Methylpropyl)-2-oxo-1-pyrrolidineacetic Acid, TRC
Produktkod. 30476284
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476284 100 mg
30464871 10 mg
30473177 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476284 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM704800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1511511-44-6
Molekylformel C10H17NO3
Molekylvikt (g/mol) 199.25
Synonym Pregabalin Impurity, 2-(4-Isobutyl-2-oxopyrrolidin-1-yl)acetic Acid
IUPAC-namn 2-[4-(2-methylpropyl)-2-oxopyrrolidin-1-yl]acetic acid
LEDER CC(C)CC1CN(CC(=O)O)C(=O)C1
Specifikationer
Kemiskt namn eller material 4-(2-Methylpropyl)-2-oxo-1-pyrrolidineacetic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C10H17NO3/c1-7(2)3-8-4-9(12)11(5-8)6-10(13)14/h7-8H,3-6H2,1-2H3,(H,13,14)
Formel vikt 199.1208