Läs mer
4-(2-Propen-1-yl)-1,2-benzenediol 1-Methanesulfonate, TRC
Produktkod. 30415483
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415483 25 mg
30418265 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30415483 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76860525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(2-Propen-1-yl)-1,2-benzenediol 1-Methanesulfonate
CAS 115234-25-8
Molekylformel C10H12O4S
Kvantitet 25 mg
Synonym 1,2-Benzenediol','4-(2-propenyl)-','1-methanesulfonate
InChI formel InChI=1S/C10H12O4S/c1-3-4-8-5-6-10(9(11)7-8)14-15(2,12)13/h3,5-7,11H,1,4H2,2H3
LEDER O=S(OC1=CC=C(CC=C)C=C1O)(C)=O
IUPAC-namn 4-allyl-2-hydroxyphenyl methanesulfonate
Molekylvikt (g/mol) 228.26
Formel vikt 228.05