missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10786285
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 127406-56-8
Melting Point 51°C to 53°C
Flash Point 113°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C12H9NO
MDL Number MFCD01863537
Quantity 5 g
Synonym 4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
InChI Key NMLYGLCBSFKJFI-UHFFFAOYSA-N
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=N1
IUPAC Name 4-pyridin-2-ylbenzaldehyde
Molecular Weight (g/mol) 183.21
PubChem CID 1515240
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-(2-Pyridyl)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-(2-Pyridinyl)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'