missing translation for 'onlineSavingsMsg'
Learn More

4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7039794
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
775351-40-1
C13H17BN4O2
272.115
MFCD12761127
ONAVJWUKLJDJAI-UHFFFAOYSA-N
5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
45785702
5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3
This item is not returnable. View return policy
775351-40-1
C13H17BN4O2
272.115
MFCD12761127
ONAVJWUKLJDJAI-UHFFFAOYSA-N
5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
45785702
5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 272.11
Percent Purity 95%
Chemical Name or Material 4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'