Läs mer
4-[(2R)-Oxiranylmethoxy]benzenepropanoic Acid Methyl Ester, TRC
Produktkod. 30423575
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423575 100 mg
30413212 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO847045100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 246219-23-8
Molekylformel C13H16O4
Molekylvikt (g/mol) 236.26
IUPAC-namn methyl 3-[4-[[(2R)-oxiran-2-yl]methoxy]phenyl]propanoate
LEDER COC(=O)CCc1ccc(OC[C@H]2CO2)cc1
Specifikationer
Kemiskt namn eller material 4-[(2R)-Oxiranylmethoxy]benzenepropanoic Acid Methyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C13H16O4/c1-15-13(14)7-4-10-2-5-11(6-3-10)16-8-12-9-17-12/h2-3,5-6,12H,4,7-9H2,1H3/t12-/m0/s1
Formel vikt 236.1049