Läs mer
4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hydrochloride, Mikromol™
Produktkod. 17991926
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991926 100 mg
17932266 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17991926 Leverantör Mikromol Leverantörsnummer MM0027.24
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hydrochloride
CAS 1956321-87-1
Analyt- eller komponentnamn 4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hydrochloride
Frakt skick Room Temperature
Molekylformel C13 H19 N O3 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Bisoprolol Fumarate Imp. L (EP), Bisoprolol Imp. L (EP), Metoprolol Succinate Imp. C (EP), Metoprolol Tartrate Imp. C (EP), Metoprolol Imp. C (EP), Metoprolol USP Related Compound C, Metoprolol USP RC C, 4-[[(2RS)-2-Hydroxy-3-(isopropylamino)propyl]oxy]benzaldehyde Hydrochloride, 4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hydrochloride, Bisoprolol Fumarate Imp. L (EP) as Hydrochloride, Metoprolol Succinate Imp. C (EP) as Hydrochloride, Metoprolol Tartrate Imp. C (EP) as Hydrochloride, Metoprolol Related Compound C as Hydrochloride, Bisoprolol Fumarate Impurity L as Hydrochloride, Metoprolol Succinate Impurity C as Hydrochloride, Metoprolol Tartrate Impurity C as Hydrochloride, Bisoprolol Impurity L as Hydrochloride, Metoprolol Impurity C as Hydrochloride
InChI formel InChI=1S/C13H19NO3.ClH/c1-10(2)14-7-12(16)9-17-13-5-3-11(8-15)4-6-13;/h3-6,8,10,12,14,16H,7,9H2,1-2H3;1H
LEDER Cl.CC(C)NCC(O)COc1ccc(C=O)cc1
IUPAC-namn 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]benzaldehyde;hydrochloride
Molekylvikt (g/mol) 273.76
Kvantitet 100 mg
Formel vikt 273.1132 g/mol