Läs mer
4-(3’,4’-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one Oxime, TRC
Produktkod. 30374867
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374867 50 mg
1 options
Produktkod. 30374867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43571050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one Oxime
CAS 152642-35-8
Molekylformel C16H13Cl2NO
Kvantitet 50 mg
Synonym 1(2H)-Naphthalenone, 4-(3,4-dichlorophenyl)-3,4-dihydro-, oxime, 4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone oxime
InChI formel InChI=1S/C16H13Cl2NO/c17-14-7-5-10(9-15(14)18)11-6-8-16(19-20)13-4-2-1-3-12(11)13/h1-5,7,9,11,20H,6,8H2/b19-16+
LEDER O\N=C\1/CCC(c2ccc(Cl)c(Cl)c2)c3ccccc13
Molekylvikt (g/mol) 306.19