missing translation for 'onlineSavingsMsg'
Learn More

4-(3,4-Difluorophenyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Catalog Number p-2700589
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175135-32-7
C9H6F2N2S
212.218
MFCD00052876
NDCSJUJQMRFHEX-UHFFFAOYSA-N
4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523
706326
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F
This item is not returnable. View return policy
175135-32-7
C9H6F2N2S
212.218
MFCD00052876
NDCSJUJQMRFHEX-UHFFFAOYSA-N
4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523
706326
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.22
Percent Purity 97%
Chemical Name or Material 4-(3, 4-Difluorophenyl)-1, 3-thiazol-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'