Läs mer
4-(3',6'-Dimethyl-3'-heptyl)phenol-13C6, TRC
Produktkod. 17992715
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17992715 1 mg -
17933785 0.5 mg 0.5mg
17981005 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17992715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4731321MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173020-38-6
Molekylformel 13C6 C9 H24 O
Molekylvikt (g/mol) 226.31
Synonym 4-(1-Ethyl-1,4-dimethylpentyl)-phenol 13C6, Phenol-1,2,3,4,5,6-13C6, 4-(1-ethyl-1,4-dimethylpentyl)-
IUPAC-namn 4-(3,6-dimethylheptan-3-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
LEDER CCC(C)(CCC(C)C)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material 4-(1-Ethyl-1,4-dimethylpentyl)-phenol 13C6
Kvantitet 1 mg
InChI formel InChI=1S/C15H24O/c1-5-15(4,11-10-12(2)3)13-6-8-14(16)9-7-13/h6-9,12,16H,5,10-11H2,1-4H3/i6+1,7+1,8+1,9+1,13+1,14+1
Formel vikt 226.2028