Läs mer
4-(3-Chloropropyl)morpholine, TRC
Produktkod. 30359619
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359619 100 mg
30356578 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC379750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7357-67-7
Molekylformel C7 H14 Cl N O
Molekylvikt (g/mol) 163.65
Synonym Morpholine, 4-(3-chloropropyl)-, 4-(3-Chloropropyl)morpholine, 1-Chloro-3-morpholinopropane, 3-(Morpholin-4-yl)propyl chloride, 3-Chloro-1-(morpholin-4-yl)propane, 3-Morpholinopropyl chloride, N-(3-Chloropropyl)morpholine, NSC 28831, NSC 38889
IUPAC-namn 4-(3-chloropropyl)morpholine
LEDER ClCCCN1CCOCC1
Specifikationer
Kemiskt namn eller material 4-(3-Chloropropyl)morpholine
Kvantitet 100 mg
InChI formel InChI=1S/C7H14ClNO/c8-2-1-3-9-4-6-10-7-5-9/h1-7H2
Formel vikt 163.0764