Läs mer
4-(3-ethoxy-4-hydroxyphenyl)butan-2-one, TRC
Produktkod. 30132477
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132477 50 mg
30012537 10 mg
30006690 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132477 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67746050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 569646-79-3
Molekylformel C12H16O3
Molekylvikt (g/mol) 208.25
LEDER CCOC1=C(C=CC(=C1)CCC(=O)C)O
Specifikationer
Kemiskt namn eller material 4-(3-ethoxy-4-hydroxyphenyl)butan-2-one
Kvantitet 50 mg
InChI formel InChI=1S/C12H16O3/c1-3-15-12-8-10(5-4-9(2)13)6-7-11(12)14/h6-8,14H,3-5H2,1-2H3
Formel vikt 208.1099