Läs mer
4-(3-Methylphenyl)-2H-pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one 1,1-Dioxide Sodium Salt (Torsemide im, TRC
Produktkod. 30483570
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30483570 50 mg
1 options
Produktkod. 30483570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32590150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(3-Methylphenyl)-2H-pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-o
CAS 72810-61-8
Molekylformel C13 H11 N3 O3 S
Kvantitet 50 mg
Synonym 2H-Pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one, 4-(3-methylphenyl)-, 1,1-dioxide, 4-(3-Methylphenyl)-2H-pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one 1,1-Dioxide, 2H-Pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one, 4-(3-methylphenyl)-, 1,1-dioxide, 4-(3-Methylphenyl)-2H-pyrido[4,3-e]-1,2,4-thiadiazin-3(4H)-one 1,1-Dioxide, Torsemide Related Compound E, Torasemide, Anhydrous Impurity A, Torasemide Impurity A
InChI formel InChI=1S/C13H11N3O3S/c1-9-3-2-4-10(7-9)16-11-5-6-14-8-12(11)20(18,19)15-13(16)17/h2-8H,1H3,(H,15,17)
LEDER Cc1cccc(c1)N2C(=O)NS(=O)(=O)c3cnccc23
IUPAC-namn 4-(3-methylphenyl)-1,1-dioxopyrido[4,3-e][1,2,4]thiadiazin-3-one
Molekylvikt (g/mol) 289.31
Formel vikt 289.0521