Learn More
[[4-[(3-Methylphenyl)amino]-3-pyridinyl]sulfonyl]carbamic Acid Ethyl Ester, TRC
Product Code. 30009038
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009038 100 mg 100mg
30137029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009038 Supplier Toronto Research Chemicals Supplier No. TRCM320685100MG
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72810-57-2
Molecular Formula C15 H17 N3 O4 S
Molecular Weight (g/mol) 335.38
Synonym Ethyl [[4-[(3-Methylphenyl)amino]pyridin-3-yl]sulfonyl]carbamate, Torasemide, Anhydrous Imp. E (EP)
IUPAC Name ethyl N-[4-(3-methylanilino)pyridin-3-yl]sulfonylcarbamate
SMILES CCOC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H17N3O4S/c1-3-22-15(19)18-23(20,21)14-10-16-8-7-13(14)17-12-6-4-5-11(2)9-12/h4-10H,3H2,1-2H3,(H,16,17)(H,18,19)
Formula Weight 335.094
Chemical Name or Material ethyl [(4-[(3-methylphenyl)amino]pyridin-3-yl)sulfonyl]carbamate