Läs mer
4,4'-(1,3,3-Trimethyl-1-propene-1,3-diyl)bisphenol, TRC
Produktkod. 30454637
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30454637 2.5 mg 2.5mg
30456224 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454637 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7966552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57244-54-9
Molekylformel C18H20O2
Molekylvikt (g/mol) 268.35
LEDER CC(=CC(C)(C)C1=CC=C(C=C1)O)C2=CC=C(C=C2)O
Specifikationer
Kemiskt namn eller material 4,4'-(1,3,3-Trimethyl-1-propene-1,3-diyl)bisphenol
Kvantitet 2.5 mg
InChI formel InChI=1S/C18H20O2/c1-13(14-4-8-16(19)9-5-14)12-18(2,3)15-6-10-17(20)11-7-15/h4-12,19-20H,1-3H3
Formel vikt 268.1463