Läs mer
4-[4-(3-Chloropropyl)-1-piperazinyl]-2-(1,1-dimethylethyl)-6-(trifluoromethyl)pyrimidine, TRC
Produktkod. 30359492
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359492 50 mg
30354390 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359492 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38121550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[4-(3-Chloropropyl)-1-piperazinyl]-2-(1,1-dimethylethyl)-6-(trifluoromethyl)pyrimidine
CAS 219600-00-7
Molekylformel C16H24ClF3N4
Kvantitet 50 mg
Synonym 2-tert-Butyl-4-[4-(3-chloropropyl)piperazin-1-yl]-6-(trifluoromethyl)pyrimidine
InChI formel InChI=1S/C16H24ClF3N4/c1-15(2,3)14-21-12(16(18,19)20)11-13(22-14)24-9-7-23(8-10-24)6-4-5-17/h11H,4-10H2,1-3H3
LEDER CC(C)(C)c1nc(cc(n1)C(F)(F)F)N2CCN(CCCCl)CC2
IUPAC-namn 2-tert-butyl-4-[4-(3-chloropropyl)piperazin-1-yl]-6-(trifluoromethyl)pyrimidine
Molekylvikt (g/mol) 364.84
Formel vikt 364.1642