Läs mer
4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one, Mikromol™
Produktkod. 30462909
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462909 4 x 25 mg
30050118 25 mg
30052123 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462909 Leverantör Mikromol Leverantörsnummer MM0258.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one
CAS 74855-91-7
Analyt- eller komponentnamn 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Frakt skick Room Temperature
Molekylformel C34 H36 Cl2 N8 O4
Certifikat och efterlevnad ISO 9001
Synonym 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one, Itraconazole Imp. C (EP), Itraconazole Impurity C
InChI formel InChI=1S/C34H36Cl2N8O4/c1-2-13-44-33(45)43(24-39-44)28-6-4-26(5-7-28)40-14-16-41(17-15-40)27-8-10-29(11-9-27)46-19-30-20-47-34(48-30,21-42-23-37-22-38-42)31-12-3-25(35)18-32(31)36/h3-12,18,22-24,30H,2,13-17,19-21H2,1H3/t30-,34-/m0/s1
Lösningstyp Neat
LEDER CCCN1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC[C@H]5CO[C@@](Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4
IUPAC-namn 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-1,2,4-triazol-3-one
Molekylvikt (g/mol) 691.61
Kvantitet 4 x 25 mg
Formel vikt 690.2237 g/mol