Läs mer
4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-Alpha,Alpha-dimethyl-benzeneacetic Acid, TRC
Produktkod. 30369160
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369160 100 mg
30371701 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30369160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD492020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1187954-57-9
Molekylformel C32 H37 N O3
Molekylvikt (g/mol) 483.64
Synonym Benzeneacetic acid, 4-[4-[4-(diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-α,α-dimethyl-, 2-[4-[(1RS)-4-[4-(Diphenylmethyl-idene)piperidin-1-yl]-1-hydroxybutyl]phenyl]-2-methylpropanoic Acid
IUPAC-namn 2-[4-[4-(4-benzhydrylidenepiperidin-1-yl)-1-hydroxybutyl]phenyl]-2-methylpropanoic acid
LEDER CC(C)(C(=O)O)c1ccc(cc1)C(O)CCCN2CCC(=C(c3ccccc3)c4ccccc4)CC2
Specifikationer
Kemiskt namn eller material 4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-alpha,alpha-dimethyl-benzeneacetic acid
Kvantitet 100 mg
InChI formel InChI=1S/C32H37NO3/c1-32(2,31(35)36)28-17-15-24(16-18-28)29(34)14-9-21-33-22-19-27(20-23-33)30(25-10-5-3-6-11-25)26-12-7-4-8-13-26/h3-8,10-13,15-18,29,34H,9,14,19-23H2,1-2H3,(H,35,36)
Formel vikt 483.2773