Läs mer
4,4',4' '-Tri-tert-butyl-2,2':6',2' '-terpyridine, TRC
Produktkod. 30410353
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410353 100 mg
30422235 50 mg
30428437 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT115090100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115091-29-7
Molekylformel C27H35N3
Molekylvikt (g/mol) 401.59
Synonym 4,4',4' '-Tris(1,1-dimethylethyl)-2,2':6',2' '-terpyridine, 2,6-Bis[4-(tert-butyl)pyridin-2-yl)-4-(tert-butyl)pyridine
IUPAC-namn 4-tert-butyl-2,6-bis(4-tert-butylpyridin-2-yl)pyridine
LEDER CC(C)(C)c1ccnc(c1)c2cc(cc(n2)c3cc(ccn3)C(C)(C)C)C(C)(C)C
Specifikationer
Kemiskt namn eller material 4,4',4' '-Tri-tert-butyl-2,2':6',2' '-terpyridine
Kvantitet 100 mg
InChI formel InChI=1S/C27H35N3/c1-25(2,3)18-10-12-28-21(14-18)23-16-20(27(7,8)9)17-24(30-23)22-15-19(11-13-29-22)26(4,5)6/h10-17H,1-9H3
Formel vikt 401.2831