Läs mer
4,4,4-Trifluoro-1-(3-methylphenyl)-1,3-butanedione, TRC
Produktkod. 30451310
Change view
Kvantitet:
500 mg
Product Code. Kvantitet
30451310 500 mg
1 options
Product Code. 30451310 Supplier Toronto Research Chemicals Supplier No. TRCT791618500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4,4,4-Trifluoro-1-(3-methylphenyl)-1,3-butanedione
CAS 53764-99-1
Molekylformel C11H9F3O2
Kvantitet 500 mg
Synonym 1,1,1-Trifluoro-4-(3'-methylphenyl)butane-2,4-dione
InChI formel InChI=1S/C11H9F3O2/c1-7-3-2-4-8(5-7)9(15)6-10(16)11(12,13)14/h2-5H,6H2,1H3
LEDER Cc1cccc(c1)C(=O)CC(=O)C(F)(F)F
IUPAC-namn 4,4,4-trifluoro-1-(3-methylphenyl)butane-1,3-dione
Molekylvikt (g/mol) 230.18
Formel vikt 230.0555