Learn More
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 326-06-7 |
| Molecular Formula | C10H7F3O2 |
| MDL Number | MFCD00000425 |
| Quantity | 500 g |
| SMILES | FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1 |
| Molecular Weight (g/mol) | 216.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 4,4,4-Trifluoro-1-phenyl-1,3-butanedione |