Läs mer
4,4,4-Trifluoro-2-(aminomethylphenyl)methylene-3-oxo-butanoic Acid Ethyl Ester, TRC
Produktkod. 30449043
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449043 500 mg
30453876 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT789900500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4,4-Trifluoro-2-(aminomethylphenyl)methylene-3-oxo-butanoic Acid Ethyl Ester
CAS 1798421-25-6
Molekylformel C14H14F3NO3
Kvantitet 500 mg
Synonym 2-[Amino-(4-methylphenyl)]methylene-4,4,4-trifluoro-3-oxo-butanoic Acid Ethyl Ester
InChI formel InChI=1S/C14H14F3NO3/c1-3-21-13(20)10(12(19)14(15,16)17)11(18)9-6-4-8(2)5-7-9/h4-7H,3,18H2,1-2H3/b11-10-
LEDER CCOC(=O)\C(=C(/N)\c1ccc(C)cc1)\C(=O)C(F)(F)F
IUPAC-namn ethyl (2Z)-2-[amino-(4-methylphenyl)methylidene]-4,4,4-trifluoro-3-oxobutanoate
Molekylvikt (g/mol) 301.26
Formel vikt 301.0926