missing translation for 'onlineSavingsMsg'
Learn More

4,4',4''-Tris[2-naphthyl(phenyl)amino]triphenylamine, 98%, Alfa Aesar™

Brand:  Alfa Aesar H64857.MD

Additional Details : CAS Number : 185690-41-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15499196

  • 1358.32 SEK / 250mg


Estimated Shipment date
from Supplier 15-07-2019
Add to basket

Specifications

Chemical Identifiers

185690-41-9
897.138
KDOQMLIRFUVJNT-UHFFFAOYSA-N
16184079
C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC6=CC=CC=C6C=C5)C7=CC=C(C=C7)N(C8=CC=CC=C8)C9=CC1=CC=CC=C1C=C9)C1=CC2=CC=CC=C2C=C1
C66H48N4
MFCD01310662
n1-naphthalen-2-yl-n4,n4-bis 4-naphthalen-2-yl phenyl amino phenyl-n1-phenylbenzene-1,4-diamine, 4,4',4-tris 2-naphthyl phenyl amino triphenylamine, tris 4-2-naphthylphenylamino phenyl amine, 4,4',4-tris n-naphth-2-yl-n-phenylamino triphenylamine, 4,4',4-tris n-2-naphthyl-n-phenyl-amino-triphenylamine, n1,n1-bis 4-naphthalen-2-yl phenyl amino phenyl-n4-naphthalen-2-yl-n4-phenylbenzene-1,4-diamine, 2t-nata, 4,4',4-tris 2-naph, 4,4,4-tris 2-naphthyl phenyl amino triphenylamine, 4,4',4'-tris 2-naphthyl phenyl amino triphenylamine
4-N-naphthalen-2-yl-1-N,1-N-bis[4-(N-naphthalen-2-ylanilino)phenyl]-4-N-phenylbenzene-1,4-diamine

Specifications

4,4',4''-Tris[2-naphthyl(phenyl)amino]triphenylamine
250mg
C66H48N4
14393968
KDOQMLIRFUVJNT-UHFFFAOYSA-N
4-N-naphthalen-2-yl-1-N,1-N-bis[4-(N-naphthalen-2-ylanilino)phenyl]-4-N-phenylbenzene-1,4-diamine
16184079
Solid
241°C to 248°C
185690-41-9
MFCD01310662
n1-naphthalen-2-yl-n4,n4-bis 4-naphthalen-2-yl phenyl amino phenyl-n1-phenylbenzene-1,4-diamine, 4,4',4-tris 2-naphthyl phenyl amino triphenylamine, tris 4-2-naphthylphenylamino phenyl amine, 4,4',4-tris n-naphth-2-yl-n-phenylamino triphenylamine, 4,4',4-tris n-2-naphthyl-n-phenyl-amino-triphenylamine, n1,n1-bis 4-naphthalen-2-yl phenyl amino phenyl-n4-naphthalen-2-yl-n4-phenylbenzene-1,4-diamine, 2t-nata, 4,4',4-tris 2-naph, 4,4,4-tris 2-naphthyl phenyl amino triphenylamine, 4,4',4'-tris 2-naphthyl phenyl amino triphenylamine
C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC6=CC=CC=C6C=C5)C7=CC=C(C=C7)N(C8=CC=CC=C8)C9=CC1=CC=CC=C1C=C9)C1=CC2=CC=CC=C2C=C1
897.138
897.14
98%
Videos
SDS