Läs mer
4,4,5,5,6,6,7,7,7-Nonafluoroheptan-1-ol, TRC
Produktkod. 30338102
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30338102 5 g
1 options
Produktkod. 30338102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5042805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4,5,5,6,6,7,7,7-Nonafluoroheptan-1-ol
CAS 83310-97-8
Molekylformel C7H7OF9
Kvantitet 5 g
Synonym 1H,1H,2H,2H,3H,3H-Perfluoroheptanol, 1-Heptanol, 4,4,5,5,6,6,7,7,7-nonafluoro- (9CI, ACI), 4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol (ACI), 1H,1H,2H,2H,3H,3H-Perfluoro-1-heptanol, 3-(Perfluorobutyl)-1-propanol, 3-(Perfluorobutyl)propanol, 4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol, 1H,1H,2H,2H,3H,3H-Perfluoro-1-heptanol, 3-(Perfluorobutyl)-1-propanol, 3-(Perfluorobutyl)propanol
InChI formel InChI=1S/C7H7F9O/c8-4(9,2-1-3-17)5(10,11)6(12,13)7(14,15)16/h17H,1-3H2
LEDER OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 4,4,5,5,6,6,7,7,7-nonafluoroheptan-1-ol
Molekylvikt (g/mol) 278.11
Formel vikt 278.0353